Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tlr13 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117183 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-183 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-183 Supplier Abnova Corporation Supplier No. PAB14610

Specifications

Antigen Tlr13
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation In serum (50% glycerol, 0.025% sodium azide)
Gene Symbols Tlr13
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 817-829 of mouse Tlr13.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 279572
Target Species Mouse, Rat
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.