Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR3 Rabbit anti-Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89114157 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-157 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-157 Supplier Abnova Corporation Supplier No. PAB0341
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Tlr3.

Sequence: SIHKIKSNPFKNQKNL, CRRFKVHHAVQQAIEQN, CILNWPVQKERINAFH

Specifications

Antigen TLR3
Applications Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Tlr3.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene Tlr3
Gene Alias AI957183
Gene Symbols Tlr3
Host Species Rabbit
Immunogen A mixture of synthetic peptides corresponding to amino acids 135-150, 828-844 and 876-891 of mouse Tlr3.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 142980
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.