Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TM9SF4 Rabbit anti-Dog, Human, Monkey, Pig, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89129697 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-697 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-697 Supplier Abnova Corporation Supplier No. PAB28190
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of TM9SF4.

Specifications

Antigen TM9SF4
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of TM9SF4.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/ml)
Formulation In PBS (0.09% sodium azide)
Gene TM9SF4
Gene Alias KIAA0255/dJ836N17.2
Gene Symbols TM9SF4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 14 amino acids at N-terminus region of human TM9SF4.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9777
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Target Species Canine, Human, Monkey, Pig, Rabbit
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.