Learn More
TMED10 Rabbit anti-Bovine, Human, Mouse, Primates, Rabbit, Rat, Biotin, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TMED10.
Supplier: Abnova Corporation PAB15140
Description
This gene is a member of the EMP24/GP25L/p24 family and encodes a protein with a GOLD domain. This type I membrane protein is localized to the plasma membrane and golgi cisternae and is involved in vesicular protein trafficking. The protein is also a member of a heteromeric secretase complex and regulates the complex's gamma-secretase activity without affecting its epsilon-secretase activity. Mutations in this gene have been associated with early-onset familial Alzheimer's disease. This gene has a pseudogene on chromosome 8. [provided by RefSeq]
Specifications
TMED10 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of TMED10. | |
In PBS | |
P24(DELTA)/S31I125/S31III125/TMP21/Tmp-21-I | |
Rabbit | |
100 μL | |
Primary | |
This antibody is useful for Western blot analysis where a band is seen at ∼21 KDa. | |
Store at 4°C. Do not freeze. |
Immunohistochemistry (PFA fixed), Western Blot | |
Biotin | |
Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user. | |
TMED10 | |
TMED10 | |
A synthetic peptide corresponding to amino acids 100-200 of human TMED10. | |
RUO | |
10972 | |
Bovine, Human, Mouse, Primate, Rabbit, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only