Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMED10 Rabbit anti-Bovine, Human, Mouse, Primates, Rabbit, Rat, DyLight 488, Polyclonal Antibody, Abnova™

Catalog No. 89117466 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-466 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-466 Supplier Abnova Corporation Supplier No. PAB15138
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of TMED10.

This gene is a member of the EMP24/GP25L/p24 family and encodes a protein with a GOLD domain. This type I membrane protein is localized to the plasma membrane and golgi cisternae and is involved in vesicular protein trafficking. The protein is also a member of a heteromeric secretase complex and regulates the complex's gamma-secretase activity without affecting its epsilon-secretase activity. Mutations in this gene have been associated with early-onset familial Alzheimer's disease. This gene has a pseudogene on chromosome 8. [provided by RefSeq]

Specifications

Antigen TMED10
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate DyLight 488
Description Rabbit polyclonal antibody raised against synthetic peptide of TMED10.
Dilution Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium borate
Gene TMED10
Gene Alias P24(DELTA)/S31I125/S31III125/TMP21/Tmp-21-I
Gene Symbols TMED10
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 100-200 of human TMED10.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10972
Test Specificity This antibody is useful for Western blot analysis where a band is seen at ∼21 KDa.
Target Species Bovine, Human, Mouse, Primate, Rabbit, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.