Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM204 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant TMEM204.
Supplier: Abnova Corporation PAB20687
Description
C16ORF30 plays a role in cell adhesion and cellular permeability at adherens junctions (Kearsey et al., 2004 [PubMed 15206924]).[supplied by OMIM
Sequence: HKREDCMAPRVIVISRSLTARFRRGLDNDYVESPCSpecifications
TMEM204 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant TMEM204. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
C16orf30/CLP24/FLJ20898/MGC111564 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
79652 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
TMEM204 | |
TMEM204 | |
Recombinant protein corresponding to amino acids of human TMEM204. | |
100 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction