Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM237, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TMEM237.
Supplier: Abnova Corporation PAB17607
Specifications
TMEM237 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of TMEM237. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
DKFZp313L091/FLJ33282 | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total ALS2CR4 protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
TMEM237 | |
TMEM237 | |
A synthetic peptide corresponding to internal of human TMEM237. | |
RUO | |
65062 | |
Human, Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction