Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFRSF10D Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114856 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-856 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-856 Supplier Abnova Corporation Supplier No. PAB1053

Rabbit polyclonal antibody raised against synthetic peptide of TNFRSF10D.

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor contains an extracellular TRAIL-binding domain, a transmembrane domain, and a truncated cytoplamic death domain. This receptor does not induce apoptosis, and has been shown to play an inhibitory role in TRAIL-induced cell apoptosis. [provided by RefSeq

Specifications

Antigen TNFRSF10D
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description tumor necrosis factor receptor superfamily, member 10d, decoy with truncated death domain
Formulation In PBS (0.02% sodium azide)
Gene TNFRSF10D
Gene Alias CD264, DCR2, TRAILR4, TRUNDD
Gene Symbols TNFRSF10D
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-terminus of human TNFRSF10D.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8793
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.