Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFRSF21 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89116837 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-837 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-837 Supplier Abnova Corporation Supplier No. PAB13779
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant TNFRSF21.

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor has been shown to activate NF-kappaB and MAPK8/JNK, and induce cell apoptosis. Through its death domain, this receptor interacts with TRADD protein, which is known to serve as an adaptor that mediates signal transduction of TNF-receptors. Knockout studies in mice suggested that this gene plays a role in T-helper cell activation, and may be involved in inflammation and immune regulation. [provided by RefSeq

Specifications

Antigen TNFRSF21
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant TNFRSF21.
Dilution Western Blot (1 ug/mL) Immunohistochemistry (3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene TNFRSF21
Gene Alias BM-018/DR6/MGC31965
Gene Symbols TNFRSF21
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 372-665 of human TNFRSF21.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27242
Test Specificity Recognizes human DR6.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.