Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFRSF6B Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89126640 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-126-640 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-126-640 Supplier Abnova Corporation Supplier No. PAB24965
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of TNFRSF6B.

This gene belongs to the tumor necrosis factor receptor superfamily. The encoded protein is postulated to play a regulatory role in suppressing FasL- and LIGHT-mediated cell death. It acts as a decoy receptor that competes with death receptors for ligand binding. Overexpression of this gene has been noted in gastrointestinal tract tumors, and it is located in a gene-rich cluster on chromosome 20, with other potentially tumor-related genes. Two transcript variants encoding the same isoform, but differing in the 5' UTR, have been observed for this gene. [provided by RefSeq]

Specifications

Antigen TNFRSF6B
Applications Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of TNFRSF6B.
Dilution Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.05% sodium azide)
Gene TNFRSF6B
Gene Alias DCR3/DJ583P15.1.1/M68/TR6
Gene Symbols TNFRSF6B
Host Species Rabbit
Immunogen A synthetic peptide corresponding to TNFRSF6B.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8771
Test Specificity TNFRSF6B polyclonal antibody detects endogenous levels of TNFRSF6B protein.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.