Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFSF11 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89116745 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-745 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-745 Supplier Abnova Corporation Supplier No. PAB13566

Rabbit polyclonal antibody raised against recombinant TNFSF11.

This gene encodes a member of the tumor necrosis factor (TNF) cytokine family which is a ligand for osteoprotegerin and functions as a key factor for osteoclast differentiation and activation. This protein was shown to be a dentritic cell survival factor and is involved in the regulation of T cell-dependent immune response. T cell activation was reported to induce expression of this gene and lead to an increase of osteoclastogenesis and bone loss. This protein was shown to activate antiapoptotic kinase AKT/PKB through a signaling complex involving SRC kinase and tumor necrosis factor receptor-associated factor (TRAF) 6, which indicated this protein may have a role in the regulation of cell apoptosis. Targeted disruption of the related gene in mice led to severe osteopetrosis and a lack of osteoclasts. The deficient mice exhibited defects in early differentiation of T and B lymphocytes, and failed to form lobulo-alveolar mammary structures during pregnancy. Two alternatively spliced transcript variants have been found. [provided by RefSeq

Specifications

Antigen TNFSF11
Applications ELISA, Flow Cytometry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant TNFSF11.
Dilution ELISA (1:2000-1:10000) Flow Cytometry (1:500-1:1000) Western Blot (1:2000-1:5000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Gene TNFSF11
Gene Alias CD254/ODF/OPGL/OPTB2/RANKL/TRANCE/hRANKL2/sOdf
Gene Symbols TNFSF11
Host Species Rabbit
Immunogen Recombinant protein corresponding to human TNFSF11.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8600
Test Specificity Recognizes human RANKL. Detects a band of ∼35 KDa by Western blot.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.