Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFSF12 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89116221 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-221 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-221 Supplier Abnova Corporation Supplier No. PAB12960
Add to Cart
Add to Cart

Rabbit Polyclonal antibody raised against recombinant TNFSF12.

The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This protein is a ligand for the FN14/TWEAKR receptor. This cytokine has overlapping signaling functions with TNF, but displays a much wider tissue distribution. This cytokine can induce apoptosis via multiple pathways of cell death in a cell type-specific manner. This cytokine is also found to promote proliferation and migration of endothelial cells, and thus acts as a regulator of angiogenesis. Some transcripts that skip the last exon of this gene and continue with the second exon of the neighboring TNFSF13 gene have been identified; such read-through transcripts are contained in GeneID 407977, TNFSF12-TNFSF13. [provided by RefSeq

Specifications

Antigen TNFSF12
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit Polyclonal antibody raised against recombinant TNFSF12.
Dilution Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene TNFSF12
Gene Alias APO3L/DR3LG/MGC129581/MGC20669/TWEAK
Gene Symbols TNFSF12
Host Species Rabbit
Immunogen Recombinant protein corresponding to human TNFSF12.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8742
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.