Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNRC4, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89123623 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-623 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-623 Supplier Abnova Corporation Supplier No. PAB21916
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant TNRC4.

Members of the CELF/BRUNOL protein family contain two N-terminal RNA recognition motif (RRM) domains, one C-terminal RRM domain, and a divergent segment of 160-230 aa between the second and third RRM domains. Members of this protein family regulate pre-mRNA alternative splicing and may also be involved in mRNA editing, and translation. While several transcript variants may exist for this gene, the full-length nature of only one has been biologically validated to date. [provided by RefSeq

Sequence: PVSAIPAALGVNGYSPVPTQPTGQPAPDALYPNGVHPYPAQSPAAPVDPLQQAYAGMQHYTAAYPAAYSLVAPAFPQPPALVAQQPPPPPQQQQQQQQQQQQQQQREGPDGCNIFIYHLPQEFTDSEILQMFV

Specifications

Antigen TNRC4
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant TNRC4.
Dilution Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene TNRC4
Gene Alias BRUNOL1/CAGH4/CELF3/ERDA4/MGC57297
Gene Symbols TNRC4
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human TNRC4.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11189
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.