Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-TopBP1, Polyclonal,

Catalog No. ABE1463MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1463MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1463MI Supplier MilliporeSigma™ Supplier No. ABE1463
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects TopBP1 in Human, Mouse samples, and it is validated for Immunocytochemistry, Western Blotting

DNA topoisomerase 2-binding protein 1 (UniProt Q92547), also known as DNA topoisomerase II-binding protein 1, DNA topoisomerase II-beta-binding protein 1, TopBP1) is encoded by the TOPBP1 gene in human. TopBP1 plays crucial roles in the maintenance of genomic integrity by participating in DNA damage repair and cell cycle checkpoints regulation. Specifically, TopBP1 participates in the loading of Cdc45 for the assembly of the preinitiation complex necessary for chromosomal DNA replication. TopBP1 is also essential for the activation of cyclin E/CDK2 and Cdc7/Dbf4 kinases, as well as DNA polymerase chromatin loading. In response to replication fork stalling or DNA damage, TopBP1 mediates the activation of ATR-ATRIP complex, leading to the phosphorylation of ATR substrates, such as Chk1, Nbs1, Smc1, and H2AX. In addition, association of TopBP1 with Nbs1 is involved in homologous recombination repair upon double-stranded DNA break. TopBP1 also binds E2F1 and p53 and suppresses the pro-apoptotic activity of p53 and E2F1 in cancer cells. TopBP1 knockdown in 3T3 and murine embryonic fibroblasts induces senescence as a result of cellular DNA breakage and cell cycle arrest.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TopBP1
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Formulation Rabbit polyclonal serum with 0.05% sodium azide.
Gene Accession No. Q92547
Gene Symbols TOPBP1; KIAA0259
Host Species Rabbit
Immunogen Recombinant protein corresponding to human TopBP1.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_008958
Target Species Human, Mouse
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.