Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TP53BP1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89115881 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-881 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-881 Supplier Abnova Corporation Supplier No. PAB12507
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant TP53BP1.

Specifications

Antigen TP53BP1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant TP53BP1.
Dilution Immunocytochemistry (1:200-1:1000) Immunofluorescence (1:50-1:200) Western Blot (1:2000-1:10000) The optimal working dilution should be determined by the end user.
Formulation In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide)
Gene TP53BP1
Gene Alias 53BP1/FLJ41424/MGC138366/p202
Gene Symbols TP53BP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to human TP53BP1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7158
Test Specificity This antibody is specific to human 53BP1 protein.
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.