Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TP53INP1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114028 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-028 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-028 Supplier Abnova Corporation Supplier No. PAB0205
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of TP53INP1.

Specifications

Antigen TP53INP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of TP53INP1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% gelatin, 0.02% sodium azide)
Gene TP53INP1
Gene Alias DKFZp434M1317/FLJ22139/SIP/TP53DINP1/TP53INP1A/TP53INP1B/Teap/p53DINP1
Gene Symbols TP53INP1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human TP53INP1.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94241
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.