Learn More
TRAF2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TRAF2 .
Supplier: Abnova Corporation PAB16905
Description
The protein encoded by this gene is a member of the TNF receptor associated factor (TRAF) protein family. TRAF proteins associate with, and mediate the signal transduction from members of the TNF receptor superfamily. This protein directly interacts with TNF receptors, and forms a heterodimeric complex with TRAF1. This protein is required for TNF-alpha-mediated activation of MAPK8/JNK and NF-kappaB. The protein complex formed by this protein and TRAF1 interacts with the inhibitor-of-apoptosis proteins (IAPs), and functions as a mediator of the anti-apoptotic signals from TNF receptors. The interaction of this protein with TRADD, a TNF receptor associated apoptotic signal transducer, ensures the recruitment of IAPs for the direct inhibition of caspase activation. BIRC2/c-IAP1, an apoptosis inhibitor possessing ubiquitin ligase activity, can unbiquitinate and induce the degradation of this protein, and thus potentiate TNF-induced apoptosis. Multiple alternatively spliced transcript variants have been found for this gene, but the biological validity of only one transcript has been determined. [provided by RefSeq
Specifications
TRAF2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of TRAF2 . | |
In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
MGC:45012/TRAP/TRAP3 | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
ELISA, Immunofluorescence, Western Blot | |
Unconjugated | |
ELISA (1:60000-1:250000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
TRAF2 | |
TRAF2 | |
A synthetic peptide corresponding to internal region of human TRAF2 . | |
RUO | |
7186 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only