Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRHR Rabbit anti-Chimpanzee, Dog, Human, Monkey, Polyclonal Antibody, Abnova™

Catalog No. 89114133 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-133 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-133 Supplier Abnova Corporation Supplier No. PAB0315
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant TRHR.

Thyrotropin-releasing hormone (TRH; MIM 275120), a small neuropeptide, is widely distributed throughout the central and peripheral nervous system as well as in extraneural tissues. The peptide is synthesized in the hypothalamus and transported by the portal vascular system to the anterior pituitary where it acts on thyrotropic and lactotropic cells to promote secretion of TSH and prolactin, respectively. Thyrotropin-releasing hormone receptor is a G protein-coupled receptor that activates the inositol phospholipid-calcium-protein kinase C transduction pathway upon the binding of TRH. The TRHR gene is expressed in the thyrotrope cells of the anterior pituitary.[supplied by OMIM

Specifications

Antigen TRHR
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant TRHR.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% gelatin, 0.05% sodium azide)
Gene TRHR
Gene Alias MGC141920
Gene Symbols TRHR
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 350-400 of human TRHR.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7201
Target Species Canine, Chimpanzee, Human, Monkey
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.