Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-TRIM11, Polyclonal,

Catalog No. ABC926MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC926MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC926MI Supplier MilliporeSigma™ Supplier No. ABC926
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects TRIM11 in Human, Mouse samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

E3 ubiquitin-protein ligase TRIM11 (UniProt Q96F44; also known as Protein BIA1, RING finger protein 92, Tripartite motif-containing 11, Tripartite motif-containing protein 11) is encoded by the TRIM11 (also known as BIA1, RNF92) gene (Gene ID 81559) in human. TRIM11 belongs to a family of more than 70 TRIM/RBCC proteins characterized by their shared tripartite motif structure, also known as the RBCC (RING finger, B-box, and coiled-coil) domain. The B-box and coiled coil regions are believed to participate in protein-protein interactions and formation of macromolecular complexes. TRIM proteins are involved in diverse cellular processes, including cell proliferation, differentiation, oncogenesis, and apoptosis. The presence of a RING domain suggests that TRIM proteins function as E3 ubiquitin ligases. The neuroprotective peptide humanin, activator-recruited cofactor (ARC) 105-kDa component (ARC105), the transcription factors Pax6 and PHOX2B are all known subrates targeted by TRIM11-mediated degradation through the ubiquitin-proteasome system. In addition, TRIM11 is also reported to interfere with the early steps of HIV-1 replication in host cells in a RING domain-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM11
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q96F44
Gene Symbols TRIM11; RNF92
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human TRIM11.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_660215
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.