Learn More
TRIM25 (phospho S100), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of TRIM25.
Supplier: Abnova Corporation PAB15930
Description
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to the cytoplasm. The presence of potential DNA-binding and dimerization-transactivation domains suggests that this protein may act as a transcription factor, similar to several other members of the TRIM family. Expression of the gene is upregulated in response to estrogen, and it is thought to mediate estrogen actions in breast cancer as a primary response gene. [provided by RefSeq
Specifications
TRIM25 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of TRIM25. | |
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
EFP/RNF147/Z147/ZNF147 | |
Rabbit | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
TRIM25 | |
TRIM25 | |
Synthetic phosphopeptide corresponding to residues surrounding S100 of human TRIM25. | |
RUO | |
7706 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only