Learn More
TRIM29 Rabbit anti-Bovine, Chimpanzee, Horse, Human, Macaque, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant TRIM29.
Supplier: Abnova Corporation PAB9976
Description
The protein encoded by this gene belongs to the TRIM protein family. It has multiple zinc finger motifs and a leucine zipper motif. It has been proposed to form homo- or heterodimers which are involved in nucleic acid binding. Thus, it may act as a transcriptional regulatory factor involved in carcinogenesis and/or differentiation. It may also function in the suppression of radiosensitivity since it is associated with ataxia telangiectasia phenotype. [provided by RefSeq
Specifications
TRIM29 | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant TRIM29. | |
In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
ATDC/FLJ36085 | |
Rabbit | |
100 μg | |
Primary | |
This antibody reacts with over-expressed, acetylated and non-acetylated (at K116) ATDC protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:100000) Western Blot (1:500-1:3000) The optimal working dilution should be determined by the end user. | |
TRIM29 | |
TRIM29 | |
Recombinant protein corresponding to internal region around K116 of human TRIM29. | |
RUO | |
23650 | |
Bovine, Chimpanzee, Equine, Human, Macaque, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only