Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-trimethyl-Histone H3 (Lys4), Alexa Fluor 647, Polyclonal,

Catalog No. 07473AF647 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 647
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
07-473-AF647 100 μL Alexa Fluor 647
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-473-AF647 Supplier MilliporeSigma™ Supplier No. 07473AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects trimethyl-Histone H3 (Lys4) in Human, Mouse samples, and it is validated for Immunocytochemistry

Histone H3 is one of the five main histone proteins that are involved in the structure of chromatin in eukaryotic cells. Histone H3 features a main globular domain and a long N-terminal tail. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine. Histone H3 is specifically phosphorylated during both mitosis and meiosis in patterns that are specifically coordinated in space and time. These phosphorylations are initiated at different phases of the cell division. Histone H3 variants (H3.1, H3.2 and H3.3) have been implicated in the epigenetic memory of cellular state. Genome-wide patterns of H3 are dependent on amino acid sequence and change with cellular differentiation at developmentally regulated loci.
TRUSTED_SUSTAINABILITY

Specifications

Antigen trimethyl-Histone H3 (Lys4)
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation Purified rabbit conjugate antibody in buffer containing PBS with 15mg/mL BSA and 0.1% sodium azide.
Gene Accession No. Q16695
Gene Symbols HIST3H3; H3FT
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues surrounding and including trimethlyated Lys 4 of Histone H3.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NM_003493
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.