Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl-Histone H3 Rabbit anti-Bovine, Chicken, Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89137530 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-530 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-530 Supplier Abnova Corporation Supplier No. PAB12741
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Trimethyl-Histone H3.

Specifications

Antigen Trimethyl-Histone H3
Applications ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Trimethyl-Histone H3.
Dilution Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (1:100) The optimal working dilution should be determined by the end user.
Formulation In TBS, pH 7.2 (BSA, 10% Proclin300)
Host Species Rabbit
Immunogen A synthetic peptide corresponding to trimethyl lysine 4 of human Histone H3.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody only recognizes ∼ 17 KDa of Trimethylated Histone H3 at K4. It does not cross react to non-methylated H3 or mono-, di-methylated Histone H3.
Target Species Bovine, Chicken, Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C or lower.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.