Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-trimethyl STAT3 (Lys180), Polyclonal,

Catalog No. ABE1397MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1397MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1397MI Supplier MilliporeSigma™ Supplier No. ABE1397
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects trimethyl STAT3 (Lys180) in Mouse samples, and it is validated for Immunoprecipitation, Western Blotting

Signal transducer and activator of transcription 3 (UniProt P40763; also known as Acute-phase response factor, DNA-binding protein APRF) is encoded by the STAT3 (also known as ADMIO, APRF, HIES) gene (Gene ID 6774) in human. STAT proteins are latent cytoplasmic transcription factors that have the dual function of signal transduction and activation of transcription. STATs are activated by tyrosine phosphorylation in response to different ligands, after which they translocate to the cell nucleus. The N-terminal region is highly homologous among the STAT proteins and surrounds a completely conserved arginine residue. STATs are a part of the JAK-STAT signaling pathway – a major pathway of the immune system. All cytokines transduce critical signals through this pathway. STAT3 is known to be activated by IFN alpha but not IFN beta. The transcription factors associated with STAT3 are cJun and cyclic AMP responsive enhancer binding protein (CREB). STAT3 gene knockout in mice is lethal at the early embryonic stage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen trimethyl STAT3 (Lys180)
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P40763
Gene Symbols STAT3; APRF
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the STAT3 K180me3 of human trimethyl STAT3 (Lys180)
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_644805
Target Species Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.