Learn More
TRMT12, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TRMT12.
Supplier: Abnova Corporation PAB19449
Description
Wybutosine (yW) is a hypermodified guanosine at the 3-prime position adjacent to the anticodon of phenylalanine tRNA that stabilizes codon-anticodon interactions during decoding on the ribosome. TRMT12 is the human homolog of a yeast gene essential for yW synthesis (Noma and Suzuki, 2006).[supplied by OMIM
Specifications
TRMT12 | |
Polyclonal | |
Unconjugated | |
Western Blot (0.5 ug/mL) Immunocytochemistry (10 ug/mL) The optimal working dilution should be determined by the end user. | |
TRMT12 | |
TRMT12 | |
A synthetic peptide corresponding to 19 amino acids near N-terminus of human TRMT12. | |
100 μg | |
Primary | |
TRMT12 antibody is predicted to not cross-react with other TYW protein family members. At least two isoforms of TYW2 are known to exist; this antibody will detect both isoforms. | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunocytochemistry, Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of TRMT12. | |
In PBS (0.02% sodium azide) | |
FLJ20772/TRM12/TYW2 | |
Rabbit | |
Peptide affinity purification | |
RUO | |
55039 | |
Human, Mouse | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only