Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trp53, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120485 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
3 X 200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-485 3 X 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-485 Supplier Abnova Corporation Supplier No. PAB18931

Rabbit polyclonal antibody raised against partial recombinant Trp53.

Specifications

Antigen Trp53
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Trp53.
Dilution Immunohistochemistry (1:500-1:1000) Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from tissue culture supernatant (0.09% sodium azide)
Gene Trp53
Gene Alias bbl/bfy/bhy/p53
Gene Symbols Trp53
Host Species Rabbit
Immunogen Recombinant protein corresponding to mouse Trp53.
Quantity 3 X 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22059
Target Species Mouse
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with sterile distilled water store at 4°C for at least two months. For long term storage store at -20°C or below. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.