Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPA1, Rabbit, DyLight 549, Polyclonal Antibody, Abnova™

Catalog No. 89117454 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-454 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-454 Supplier Abnova Corporation Supplier No. PAB15114
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of TRPA1.

The structure of the protein encoded by this gene is highly related to both the protein ankyrin and transmembrane proteins. The specific function of this protein has not yet been determined; however, studies indicate the function may involve a role in signal transduction and growth control. [provided by RefSeq

Specifications

Antigen TRPA1
Applications Immunohistochemistry, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate DyLight 549
Description Rabbit polyclonal antibody raised against synthetic peptide of TRPA1.
Dilution Immunohistochemistry (0.5-1 ug/mL) Western Blot (2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:250) The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium borate
Gene TRPA1
Gene Alias ANKTM1
Gene Symbols TRPA1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 1-100 of human TRPA1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8989
Test Specificity 69% sequence identity with mouse and rat proteins. TRPA1 This antibody can be used for Western blot analysis where a band is seen at ∼110 KDa.
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.