Learn More
TRPA1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TRPA1.
Supplier: Abnova Corporation PAB11992
Description
The structure of the protein encoded by this gene is highly related to both the protein ankyrin and transmembrane proteins. The specific function of this protein has not yet been determined; however, studies indicate the function may involve a role in signal transduction and growth control. [provided by RefSeq
Specifications
TRPA1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of TRPA1. | |
In Tris-glycine, 150mM NaCl (0.05% sodium azide) | |
ANKTM1 | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse | |
Liquid |
Immunohistochemistry, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (0.5-1 ug/mL) Western Blot (2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:250) The optimal working dilution should be determined by the end user. | |
TRPA1 | |
TRPA1 | |
A synthetic peptide corresponding to amino acids 1-100 of human TRPA1. | |
RUO | |
8989 | |
Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only