Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPM4 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89129324 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-324 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-324 Supplier Abnova Corporation Supplier No. PAB27803
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of TRPM4.

Specifications

Antigen TRPM4
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of TRPM4.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-7.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene TRPM4
Gene Alias FLJ20041/TRPM4B
Gene Symbols TRPM4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 18 amino acid at C-terminus of human TRPM4.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54795
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.