Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trpm7, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117335 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-335 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-335 Supplier Abnova Corporation Supplier No. PAB14890
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Trpm7.

Specifications

Antigen Trpm7
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Trpm7.
Dilution ELISA (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.1% proclin, 2% Block Ace)
Gene Trpm7
Gene Alias 2310022G15Rik/4833414K03Rik/5033407O22Rik/CHAK/CHAK1/Ltpr7/Ltrpc7/TRP-PLIK
Gene Symbols Trpm7
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of mouse Trpm7.
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58800
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.