Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPM8 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TRPM8.
Supplier: Abnova Corporation PAB4072
Specifications
TRPM8 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of TRPM8. | |
In PBS (0.09% sodium azide) | |
LTRPC6/MGC2849/TRPP8 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
79054 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
ELISA (1:1000) Western Blot (1:50-100) Immunohistochemistry (1:10-50) The optimal working dilution should be determined by the end user. | |
TRPM8 | |
TRPM8 | |
A synthetic peptide (conjugated with KLH) corresponding to internal region of human TRPM8. | |
400 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction