Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPV3 Rabbit anti-Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89118703 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-703 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-703 Supplier Abnova Corporation Supplier No. PAB16890
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Trpv3.

Sequence: YRPREEEAI

Specifications

Antigen TRPV3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Trpv3.
Dilution EIA (1:5000) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS
Gene Trpv3
Gene Alias 1110036I10Rik/AI644701/MGC124324/MGC124325/Nh/VRL3
Gene Symbols Trpv3
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to Trpv3.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246788
Target Species Mouse
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.