Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTYH2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89127097 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-097 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-097 Supplier Abnova Corporation Supplier No. PAB25480
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of TTYH2.

This gene encodes a member of the tweety family of proteins. Members of this family function as chloride anion channels. The encoded protein functions as a calcium(2+)-activated large conductance chloride(-) channel, and may play a role in kidney tumorigenesis. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]

Specifications

Antigen TTYH2
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of TTYH2.
Dilution Western Blot (1 ug/mL) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene TTYH2
Gene Alias C17orf29/MGC126172/MGC4837
Gene Symbols TTYH2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-terminus of human TTYH2.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94015
Test Specificity At least four isoforms of TTYH2 are known to exist; this antibody will only detect the two longest isoforms. This antibody is predicted to not cross-react with other TTYH family members.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.