Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBB, Rabbit, HRP, Polyclonal Antibody, Abnova™

Catalog No. 89117439 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-439 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-439 Supplier Abnova Corporation Supplier No. PAB15097
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of TUBB.

Specifications

Antigen TUBB
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Description Rabbit polyclonal antibody raised against synthetic peptide of TUBB.
Dilution Western Blot (1:1000-1:10000) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene TUBB
Gene Alias M40/MGC117247/MGC16435/OK/SW-cl.56/TUBB1/TUBB5
Gene Symbols TUBB
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 1-100 of human TUBB.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203068
Test Specificity This is specific to beta tubulin. This antibody can be used for Western Blot where a band at ∼50 KDa is seen.
Target Species Chicken, Human, Mouse, Pig, Rat, Xenopus species, Zebrafish
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.