Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TYRO3 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89117619 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-619 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-619 Supplier Abnova Corporation Supplier No. PAB15505
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of TYRO3.

Specifications

Antigen TYRO3
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of TYRO3.
Dilution Immunocytochemistry (1:200) Immunofluorescence (1:200) Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (30% glycerol, 0.09% sodium azide)
Gene TYRO3
Gene Alias BYK/Brt/Dtk/FLJ16467/RSE/Sky/Tif
Gene Symbols TYRO3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 100-250 of human TYRO3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7301
Test Specificity human and mouse. 100% sequence identity to human and mouse protein, 94% sequence identity to rat protein. This antibody is useful for Western blot analysis where a band can be seen at ∼96 KDa.
Target Species Human, Mouse
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.