Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquintin, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89137944 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-944 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-944 Supplier Abnova Corporation Supplier No. PAB14511
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against Ubiquintin.

Specifications

Antigen Ubiquintin
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against Ubiquintin.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:100) Immunohistochemistry (Frozen sections) (1:50-1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (1% BSA, 0.05% sodium azide)
Host Species Rabbit
Immunogen Human Ubiquitin.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bovine, Equine, Human, Mouse, Rat
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.