Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBN2 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89122768 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-122-768 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-122-768 Supplier Abnova Corporation Supplier No. PAB21277

Rabbit polyclonal antibody raised against recombinant UBN2.

Sequence: DASSLTQVTKVHQHSAVQQNYVSPLQATISKSQTNPVVKLSNNPQLSCSSSLIKTSDKPLMYRLPLSTPSPGNGSQGSHPLVSRTVPSTTT

Specifications

Antigen UBN2
Applications Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description ubinuclein 2
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene UBN2
Gene Alias FLJ25778
Gene Symbols UBN2
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human UBN2.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254048
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.