Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCHL1, Rabbit, Biotin, Polyclonal Antibody, Abnova™

Catalog No. 89117382 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-382 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-382 Supplier Abnova Corporation Supplier No. PAB14971
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of UCHL1.

The protein encoded by this gene belongs to the peptidase C12 family. This enzyme is a thiol protease that hydrolyzes a peptide bond at the C-terminal glycine of ubiquitin. This gene is specifically expressed in the neurons and in cells of the diffuse neuroendocrine system. Mutations in this gene may be associated with Parkinson disease

Specifications

Antigen UCHL1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Biotin
Description Rabbit polyclonal antibody raised against synthetic peptide of UCHL1.
Dilution Immunohistochemistry (2.5-5 ug/mL) Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene UCHL1
Gene Alias PARK5/PGP9.5/Uch-L1
Gene Symbols UCHL1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 100-200 of human UCHL1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7345
Test Specificity This antibody reacts with mouse and human UCHL1 protein. The immunogen used for this antibody production has 100% homology with monkey and 99% with rat, pig, and horse proteins. Rabbit Polyclonal to PGP9.5 - Peripheral Nerve Fiber marker This antibody is useful for Western blot and immunohistochemistry. By Western blot a band at ∼25 KDa is seen.
Target Species Bovine, Equine, Guinea Pig, Human, Mouse, Pig, Primate, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.