Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCHL1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89115883 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-883 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-883 Supplier Abnova Corporation Supplier No. PAB12509
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against full-length recombinant UCHL1.

The protein encoded by this gene belongs to the peptidase C12 family. This enzyme is a thiol protease that hydrolyzes a peptide bond at the C-terminal glycine of ubiquitin. This gene is specifically expressed in the neurons and in cells of the diffuse neuroendocrine system. Mutations in this gene may be associated with Parkinson disease

Specifications

Antigen UCHL1
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant UCHL1.
Dilution Immunofluorescence (1:500-1:1000) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation In antiserum (10mM sodium azide)
Gene UCHL1
Gene Alias PARK5/PGP9.5/Uch-L1
Gene Symbols UCHL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to full length human UCHL1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7345
Target Species Human
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.