Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCHL1 Rabbit anti-Horse, Human, Mouse, Pig, Primates, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of UCHL1.

Manufacturer:  Abnova Corporation PAB11988

Catalog No. 89-115-496


Add to cart

Description

Description

The protein encoded by this gene belongs to the peptidase C12 family. This enzyme is a thiol protease that hydrolyzes a peptide bond at the C-terminal glycine of ubiquitin. This gene is specifically expressed in the neurons and in cells of the diffuse neuroendocrine system. Mutations in this gene may be associated with Parkinson disease

Specifications

Specifications

UCHL1
Unconjugated
Western Blot (2 ug/mL) Immunohistochemistry (2.5-10 ug/mL) The optimal working dilution should be determined by the end user.
UCHL1
PARK5|PGP9.5|Uch-L1
Rabbit
100 uL
Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Equine, Human, Murine, Porcine, Primate, Rat
Immunohistochemistry, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of UCHL1.
In Tris-glycine, 150mM NaCl (0.05% sodium azide)
Liquid
UCHL1
A synthetic peptide corresponding to amino acids 200-300 of human UCHL1.
RUO
Primary
7345
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only