Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-UFD1L, Polyclonal,

Catalog No. ABN1014MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1014MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1014MI Supplier MilliporeSigma™ Supplier No. ABN1014
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects UFD1L in Human samples, and it is validated for Immunocytochemistry, Immunohistochemistry

UFD1L is an essential component of the ubiquitin dependent proteolytic pathway which degrades ubiquitin fusion proteins. UFD1L is part of the three protein complex of UFD1L, VCP and NPLOC4 that binds ubiquitinated proteins and is necessary for the export of misfolded proteins from the ER to the proteasome. The complex also regulates spindle disassembly during mitosis and plays an important role in ectoderm development as well. In the adult UFD1L is widely expressed but highest levels are in heart, skeletal muscle, and pancreas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UFD1L
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q92890
Gene Symbols UFD1L
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human UFD1L.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_005650
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.