Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ulex Europaeus Lectin 1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89138107 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-107 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-107 Supplier Abnova Corporation Supplier No. PAB15537

Rabbit polyclonal antibody raised against native Ulex Europaeus Lectin 1.

Specifications

Antigen Ulex Europaeus Lectin 1
Applications Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native Ulex Europaeus Lectin 1.
Dilution Immunofluorescence (1:500) Immunohistochemistry (Frozen sections) (1:500) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500) The optimal working dilution should be determined by the end user.
Formulation In antiserum (0.09% sodium azide)
Host Species Rabbit
Immunogen Native purified from Ulex europaeus agglutinin isolated from seeds of Ulex europaeus (gorse seed).
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody may be used for studies of the vascular structures. It reacts specifically with UEA-I bound to human endothelial cells of normal and neoplastic blood and lymphatic vessels. Its uniform reaction with vessels enables differentation between vessels and artificial slits in the tissue. Other species have not been tested.
Target Species Human, Plant
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.