Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ULP1 Rabbit anti-Yeast, Polyclonal Antibody, Abnova™

Catalog No. 89115047 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-047 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-047 Supplier Abnova Corporation Supplier No. PAB11365
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant ULP1.

Specifications

Antigen ULP1
Applications ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant ULP1.
Dilution ELISA (1:4000-1:20000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene ULP1
Gene Alias NIB1
Gene Symbols ULP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Saccharomyces cerevisiae ULP1.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 856087
Target Species Yeast
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 100μL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.