Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ulp2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114499 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-499 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-499 Supplier Abnova Corporation Supplier No. PAB0936

Rabbit polyclonal antibody raised against synthetic peptide of Ulp2.

Sequence: SNNERQSLSSGSND

Specifications

Antigen Ulp2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description SUMO deconjugating cysteine peptidase Ulp2 (predicted)
Formulation In PBS (0.02% sodium azide)
Gene ulp2
Gene Symbols ulp2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 622-635 of Schizosaccharomyces pombe Ulp2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2542268
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.