Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UPK3A, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117298 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-298 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-298 Supplier Abnova Corporation Supplier No. PAB14839

Rabbit polyclonal antibody raised against synthetic peptide of UPK3A.

Uroplakins (UPK) are integral membrane proteins that form 2-dimensional crystalline arrays termed urothelial plaques that cover more than 90% of the apical urothelial surface. The asymmetric unit membrane (AUM) forms the apical plaques of urothelium and is believed to strengthen the urothelial apical surface and prevent the cells from rupturing during bladder distention. The 4 major conserved integral membrane proteins of the AUM are UPK1A (MIM 611557), UPK1B (MIM 602380), UPK2 (MIM 611558), and UPK3A.[supplied by OMIM

Specifications

Antigen UPK3A
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Concentration Lot specific
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of UPK3A.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4 (1% BSA, 0.09% sodium azide)
Gene UPK3A
Gene Alias MGC119178/UPIII/UPIIIA/UPK3
Gene Symbols UPK3A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human UPK3A.
Purification Method Immunoaffinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7380
Test Specificity Molecular weight: 32 KDa. Epitope: C-terminus.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.