Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Urease, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native Urease.
Supplier: Abnova Corporation PAB11382
Specifications
| Urease | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against native Urease. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| Native purified Jack bean Urease. | |
| RUO | |
| Plant | |
| Lyophilized |
| ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Sandwich ELISA (1:15000-1:60000) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction