Learn More
USP16 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of USP16.
Supplier: Abnova Corporation PAB4203
Description
This gene encodes a deubiquitinating enzyme that is phosphorylated at the onset of mitosis and then dephosphorylated at the metaphase/anaphase transition. It can deubiquitinate H2A, one of two major ubiquitinated proteins of chromatin, in vitro and a mutant form of the protein was shown to block cell division. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Specifications
USP16 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of USP16. | |
In PBS (0.09% sodium azide) | |
UBP-M | |
Rabbit | |
Protein G purification | |
RUO | |
10600 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
USP16 | |
USP16 | |
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human USP16. | |
400 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only