Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vaccinia virus G9R Rabbit anti-Viruses, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Vaccinia virus G9R.
Supplier: Abnova Corporation PAB16109
Specifications
Vaccinia virus G9R | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Vaccinia virus G9R. | |
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
A synthetic peptide corresponding to N-terminus of Vaccinia virus G9R. | |
RUO | |
Virus | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
Rabbit | |
100 μg | |
Primary | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction