Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Variola virus Protein J5 Rabbit anti-Viruses, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Variola virus Protein J5.
Supplier: Abnova Corporation PAB16105
Specifications
Variola virus Protein J5 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Variola virus Protein J5. | |
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
A synthetic peptide corresponding to C-terminus of Variola virus Protein J5. | |
RUO | |
Virus | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
Rabbit | |
100 μg | |
Primary | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction