Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VDAC1 Rabbit anti-Birds, Human, Polyclonal Antibody, Abnova™

Catalog No. 89114997 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-997 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-997 Supplier Abnova Corporation Supplier No. PAB11290
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of VDAC1.

Specifications

Antigen VDAC1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of VDAC1.
Dilution ELISA (1:10000-1:40000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene VDAC1
Gene Alias MGC111064/PORIN/PORIN-31-HL
Gene Symbols VDAC1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 185-197 of human VDAC1.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7416
Test Specificity This affinity purified antibody is directed against human VDAC1/Porin1 protein. Cross reactivity will occur with all forms of VDACs including VDAC1, VDAC2 (4 isoforms) and VDAC3 (2 isoforms).
Target Species Avian, Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.